missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRDM4 Recombinant Human Monoclonal Antibody (RAB-C367)
GREENER_CHOICE

Numéro de catalogue. MA551850
Modifier l'affichage
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA551850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MA551850 Fournisseur Invitrogen™ Code fournisseur MA551850
Il en reste null

Human Recombinant Monoclonal Antibody

This antibody recognizes PRDM4 (PR domain zinc finger protein 4). It binds to a folded domain, amino acids 390-540. PRDM4 is a transcription factor involved in cell differentiation.

The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.

Spécifications

Antigen PRDM4
Applications ChIP Assay, ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RAB-C367
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene PRDM4
Gene Accession No. Q9UKN5
Gene Alias 1700031E19Rik; 2810470D21Rik; AW552272; MGC45046; PFM1; PR domain 4; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PRDM4; PR-domain zinc-finger protein PFM1; SC-1
Gene Symbols PRDM4
Host Species Human
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions.
Purification Method Metal-chelate chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11108
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype F(ab) κ
Afficher plus de résultats Afficher moins de résultats