missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel 98.0+%, TCI America™

Catalog No. P21255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21255G 5 g
P212525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21255G Supplier TCI America Supplier No. P21255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55268-74-1
Molecular Formula C19H24N2O2
Molecular Weight (g/mol) 312.413
MDL Number MFCD00058531
InChI Key FSVJFNAIGNNGKK-UHFFFAOYSA-N
Synonym praziquantel, droncit, biltricide, pyquiton, cesol, azinox, embay 8440, cutter tape tabs, prasiquantel, praziquantelum
PubChem CID 4891
IUPAC Name 2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
SMILES C1CCC(CC1)C(=O)N2CC3C4=CC=CC=C4CCN3C(=O)C2

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 312.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Praziquantel
RTECSNumber UQ4150000
TSCA No
Recommended Storage Freezer