missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014592
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014592 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014592 Supplier Merck Emd Millipore Supplier No. PHR13911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Praziquantel is at present the only drug used for the treatment of human schistosomiasis. It has a pyrazino isoquinoline ring system and is found to be very successful in animals for its anticestode and antitrematode properties. It has been identified that the voltage-gated schistosome calcium ion (Ca2+) channels serve as a target for praziquantel. Even though the commercial preparation of praziquantel comprises of equal amounts of both its 'dextro′ and 'laevo′ isomers, only the 'laevo′ form exhibits schistosomicidal activity.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 55268-74-1
Physical Form Neat
Linear Formula C19H24N2O2
MDL Number MFCD00058531
Synonym Praziquantel; 2-(Cyclohexylcarbonyl)-1,2,3,6,7-11 b-hexahydro-4 H-pyrazino[2,1-a]isoquinolin-4-one
Type Certified Reference Material
Quantity 1 g
Formula Weight 312.41
Grade Certified Reference Material