missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium ICP Standard, 1000 ppm Pr in 3% HNO3, Ricca Chemical

Catalog No. PPR1KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PPR1KN100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PPR1KN100 Supplier Ricca Chemical Company Supplier No. PPR1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12037-29-5 , 7697-37-2 , 12037-29-5

Specifications

Packaging Natural LDPE Bottle
CAS Min % 94.0, 4.36, 0.11
CAS Max % 97.0, 4.53, 0.12
Concentration or Composition (by Analyte or Components) 1mL = 1mg Pr (1, 000ppm Pr)Pr2O3 in 3% HNO3
Chemical Name or Material Praseodymium ICP Standard
ShelfLife 18 months