missing translation for 'onlineSavingsMsg'
Learn More

Pranlukast 98.0+%, TCI America™

Catalog No. P211525MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P211525MG 25 mg
P2115100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P211525MG Supplier TCI America Supplier No. P211525MG
Only null left

Chemical Identifiers

CAS 103177-37-3
Molecular Formula C27H23N5O4
Molecular Weight (g/mol) 481.512
MDL Number MFCD00864631
InChI Key NBQKINXMPLXUET-UHFFFAOYSA-N
Synonym pranlukast, azlaire, pranlukast hydrate, onon, pranlukast inn, azlaire tn, n-4-oxo-2-2h-tetrazol-5-yl chromen-8-yl-4-4-phenylbutoxy benzamide, chembl21333, ono-rs-411, 4-oxo-8-4-4-phenylbutoxy benzoylamino-2-tetrazol-5-yl-4h-1-benzopyran
PubChem CID 4887
IUPAC Name N-[4-oxo-2-(2H-tetrazol-5-yl)chromen-8-yl]-4-(4-phenylbutoxy)benzamide
SMILES C1=CC=C(C=C1)CCCCOC2=CC=C(C=C2)C(=O)NC3=CC=CC4=C3OC(=CC4=O)C5=NNN=N5

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 25 mg
Formula Weight 481.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pranlukast
RTECSNumber CV5647500
TSCA No
Recommended Storage Refrigerator