missing translation for 'onlineSavingsMsg'
Learn More

Pramipexole Dihydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013409
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013409 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013409 Supplier Merck Emd Millipore Supplier No. PHR1598500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pramipexole Dihydrochloride is a non-ergot dopamine agonist widely used for the treatment of early and advanced Parkinson′s disease (PD). It is also used to treat restless legs syndrome.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 191217-81-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H17N3 S · 2 HCl · H2O
Synonym Pramipexole dihydrochloride monohydrate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 302.26
Grade Certified Reference Material