missing translation for 'onlineSavingsMsg'
Learn More

Pramipexole Dihydrochloride Monohydrate 98.0+%, TCI America™

Catalog No. P20731G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20731G 1 g
P2073100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20731G Supplier TCI America Supplier No. P20731G
Only null left

Chemical Identifiers

CAS 191217-81-9
Molecular Formula C10H21Cl2N3OS
Molecular Weight (g/mol) 302.26
MDL Number MFCD00876894,MFCD02183927
InChI Key APVQOOKHDZVJEX-QTPLPEIMSA-N
Synonym (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride
PubChem CID 166589
ChEBI CHEBI:51147
IUPAC Name dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2

Specifications

Melting Point 290°C
Color White-Yellow
Quantity 1 g
Formula Weight 284.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pramipexole Dihydrochloride Monohydrate
RTECSNumber DL3375000
TSCA No