missing translation for 'onlineSavingsMsg'
Learn More

PPP2R5D Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Numéro de catalogue. 501559250
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
501559250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 501559250 Fournisseur Bethyl Laboratories Inc Code fournisseur A301099A

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rabbit

The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a delta isoform of the regulatory subunit B56 subfamily. Alternatively spliced transcript variants encoding different isoforms have been identified.

Spécifications

Antigène PPP2R5D
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugué Unconjugated
Formule phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gène PPP2R5D
Numéro d’acquisition du gène Q14738
Alias du gène PP2CB
Symboles de gène PPP2R5D
Espèces hôtes Rabbit
Immunogène Between 500 and 550
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 5528
Espèces cibles Human
Contenu et stockage 4° C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats