missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP2R5A Polyclonal Antibody

Description
PPP2R5A Polyclonal Antibody for Western Blot, IHC
The product of this gene belongs to the phosphatase 2A regulatory subunit B family. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B56 subfamily. Alternative transcript variants encoding distinct isoforms have been found for this gene.
Spécifications
Spécifications
| Antigène | PPP2R5A |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | PBS with 0.08% sodium azide |
| Gène | PPP2R5A |
| Numéro d’acquisition du gène | Q15172, Q6PD03 |
| Alias du gène | B56A; MGC131915; PP2A B subunit isoform B56-alpha; PP2A B subunit isoform B'-alpha; PP2A B subunit isoform PR61-alpha; PP2A B subunit isoform R5-alpha; PP2A, B subunit, B' alpha isoform; PP2A, B subunit, B56 alpha isoform; PP2A, B subunit, PR61 alpha isoform; PP2A, B subunit, R5 alpha isoform; PP2CB; PPP2R5A; PR61A; PR61alpha; protein phosphatase 2 regulatory subunit B', alpha; protein phosphatase 2 regulatory subunit B'alpha; protein phosphatase 2, regulatory subunit B (B56), alpha isoform; protein phosphatase 2, regulatory subunit B', alpha; protein phosphatase 2, regulatory subunit B', alpha isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alpha isoform; serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform |
| Symboles de gène | PPP2R5A |
| Espèces hôtes | Sheep |
| Afficher plus de résultats |