missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPOX Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa, HCT116. Predicted reactivity: Mouse (83%), Rat (81%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protoporphyrinogen oxidase, the penultimate enzyme in the heme biosynthetic pathway, catalyzes the 6-electron oxidation of protoporphyrinogen IX to form protoporphyrin IX. The PPOX protein localizes to the inner membrane of mitochondria from various tissues, including heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. Genetic deficiency of PPOX results in variegate porphyria, a low penetrance, autosomal dominant disorder characterized by cutaneous photosensitivity and/or various neurological manifestations. The rare homozygous variant of VP is characterized by severe PPOX deficiency, and results in the onset of photosensitization by porphyrins in early childhood, skeletal abnormalities of the hand, and, less constantly, short stature, mental retardation and convulsions.
Spécifications
Spécifications
| Antigen | PPOX |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.82 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PPOX |
| Gene Accession No. | P50336 |
| Gene Alias | PPO; Ppox; protoporhyrinogen oxidase; protoporphyrinogen oxidase; V290M; VP |
| Gene Symbols | PPOX |
| Afficher plus de résultats |