missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
$214.38 - $5406.21
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Material | Epoxy-coated Steel |
| Shape | Rectangular |
| Sterility | Non-sterile |
| Diameter (Metric) Opening | 16 mm |
| Catalog Number | Mfr. No. | Color | Positions | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Positions | Price | Quantity | ||||
|
22-260553
|
Bel-Art™
F187570001 |
Blue | 72 |
Each for $214.93
Case of 24 Each for $5,158.32
|
|
||||
|
14-793-232
|
N/A | Blue | 60 |
Each for $289.44
Case of 24 Each for $6,946.56
|
|
||||
|
22-260549
|
Bel-Art™
F187550001 |
Blue | 48 |
Each for $219.21
Case of 24 Each for $5,261.04
|
|
||||
|
11-391-311
|
Bel-Art™
F187590001 |
Blue | 108 |
Each for $247.28
Case of 24 Each for $5,934.72
|
|
||||
|
22-260556
|
Bel-Art™
F187590000 |
Green | 108 |
Case of 24 Each for $5,406.21
|
|
||||
|
11-391-310
|
Bel-Art™
F187590003 |
Orange | 108 |
Each for $214.38
Case of 24 Each for $5,145.12
|
|
||||
|
14-793-236
|
N/A | Blue | 150 |
Each for $256.26
Case of 12 Each for $3,075.12
|
|
||||
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
| Autoclavable | |
| Rectangular | |
| 16 mm |
| Epoxy-coated Steel | |
| Non-sterile | |
| Test Tube Rack |