missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
| Autoclavable | |
| Tubes (13 to 16 mm) | |
| 60 | |
| Yes, When Empty | |
| 8 x 5 x 2.5 in. (20.3 x 12.9 x 6.4 cm) | |
| 8 in. | |
| 2.5 in. | |
| Test Tube Rack | |
| 6.4 cm | |
| 12.9 cm |
| Blue | |
| Epoxy-coated Steel | |
| Rectangular | |
| Non-sterile | |
| 20.3 cm | |
| 5.06 in. | |
| 16 mm | |
| 3 Tiers | |
| 6 x 10 |