missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Poxygrid™ Wire Test Tube Racks

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

 View more versions of this product

Catalog No. 14-793-236



Only null left

Description

Description

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable
Specifications

Specifications

Autoclavable
Tubes (13 to 16 mm)
150
Yes, When Empty
12 x 8.25 x 2.5 in. (30.5 x 21 x 6.4 cm)
12 in.
2.5 in.
16 mm
3 Tiers
10 x 15
Blue
Epoxy-coated Steel
Rectangular
Non-sterile
305 mm
8.25 in.
150 mm
Test Tube Rack
64 mm
209 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions