missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Supplier: Bel-Art™ F187550001
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
| Autoclavable | |
| Tubes (13 to 16 mm) | |
| 48 | |
| Yes, When Empty | |
| 9.5 x 3.6 x 2.5 in. (242 x 92 x 64 mm) | |
| 9.48 in. | |
| 2.51 in. | |
| Test Tube Rack | |
| 3 Tiers | |
| 4 x 12 |
| Blue | |
| Epoxy-coated Steel | |
| Rectangular | |
| Non-sterile | |
| 242 mm | |
| 3.62 in. | |
| 16 mm | |
| 16 mm | |
| 64 mm | |
| 92 mm |