missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Poxygrid™ Biohazard Bag Holders

Catalog No. 0181510A
Change view
Click to view available options
Diameter (English):
7.3 in.
14.6 in.
Height (English):
20.5 in.
23.5 in.
29.5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Height (English)
0181510A 7.3 in. 20.5 in.
0181510C 14.6 in. 29.5 in.
0181510B 14.6 in. 23.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0181510A Supplier Bel-Art™ Supplier No. H131870000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place

  • Bright biohazard orange colored epoxy-coated steel wire
  • Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
  • Autoclavable

Specifications

Description Circular epoxy coated wire frame with easy-to-use hooked ring over a top
Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With (Application) To keep biohazard bags open and ready for use
For Use With (Equipment) 01-814-1, 01-814A/B, 01-815A, 01-826-5, 01-826A, 01-827B, 01-828B, 01-829B, 01-830B 24 H x 12 in. W biohazard bags
Material Epoxy-coated Wire
Temperature Range (English) -369°F to +250°F
Temperature Range (Metric) -223°C to +121°C
Width (English) 7.3 in.
Height (English) 20.5 in.
Height (Metric) 52 cm
Diameter (English) 7.3 in.
Diameter (Metric) 18.5 cm
Product Type Biohazard Bag Holder
Width (Metric) 18.5 cm
Show More Show Less