missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25 kg
GREENER_CHOICE

Catalog No. SH3086403
Change view
Click to view available options
Port Detail:
Powder Port: 3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Detail
SH3086403 Powder Port: 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3086403 Supplier Thermo Scientific™ Supplier No. SH30864.03

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port)

  • The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.
  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.
  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.

Specifications

Includes 1 Port (1 Powder Port)
Film Description CX3-9
Outer Packaging Packages are placed in strong cardboard cartons with product and lot identification.
Port Detail Powder Port: 3 in.
Product Line PowderTainer™ II
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Sterilization Method Non-Irradiated
Tubing Line 1 Washdown Line: N/A
Tubing Line 2 Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal
Dimensions (L x W x H) 83.3 x 59.2 cm (32.8 x 23.3 in.)
Width (English) 23.3 in.
Weight (Metric) 25 kg
Product Type Bioprocess Container for Powder
Length (Metric) 83.3 cm
Width (Metric) 59.2 cm
Show More Show Less