missing translation for 'onlineSavingsMsg'
Learn More

Potassium tris(3,5-dimethyl-1-pyrazolyl)borohydride, 99%

Catalog No. AC296020050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296020050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296020050 Supplier Thermo Scientific Chemicals Supplier No. 296020050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17567-17-8 | C15H22BKN6 | 336.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 17567-17-8
Molecular Formula C15H22BKN6
Molecular Weight (g/mol) 336.29
MDL Number MFCD00040342
InChI Key NTWZGFNSHCFHIJ-UHFFFAOYSA-N
Synonym potassium hydrotris 3,5-dimethylpyrazol-1-yl borate, potassium;tris 3,5-dimethylpyrazol-1-yl boron 1-, potassium tri 3,5-dimethyl-1-pyrazolyl borohydride, potassium tris 3,5-dimethyl-1h-pyrazol-1-yl hydrido borate 1-
IUPAC Name potassium tris(3,5-dimethyl-1H-pyrazol-1-yl)boranuide
SMILES [K+].CC1=NN([BH-](N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 287°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: reacts. Other solubilities: soluble in alchohols,thf,dmf,acetone
Formula Weight 336.28
Percent Purity 99%
Physical Form Fine Needle-like Powder
Chemical Name or Material Potassium tris(3, 5-dimethyl-1-pyrazolyl)borohydride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
In contact with water releases flammable gas.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only