missing translation for 'onlineSavingsMsg'
Learn More

Potassium trimethylacetate, 95%

Catalog No. AAH6280322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6280322 100 g
AAH6280314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6280322 Supplier Thermo Scientific Chemicals Supplier No. H6280322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 19455-23-3 | C5H9KO2 | 140.223 g/mol

Potassium trimethylacetate is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trimethylacetate is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 19455-23-3
Molecular Formula C5H9KO2
Molecular Weight (g/mol) 140.223
MDL Number MFCD00671345
InChI Key WFMNHCSATCWAAQ-UHFFFAOYSA-M
Synonym potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt
PubChem CID 23662159
IUPAC Name potassium;2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)[O-].[K+]

Specifications

Melting Point ∼200°C
Linear Formula (CH3)3CCO2K
Quantity 100 g
Formula Weight 140.22
Percent Purity 95%
Chemical Name or Material Potassium trimethylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only