missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trimethoxy(trifluoromethyl)borate 95.0+%, TCI America™

Catalog No. p-7135368
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20365G 5 g
P203625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20365G Supplier TCI America Supplier No. P20365G

Chemical Identifiers

CAS 626232-27-7
Molecular Formula C4H9BF3KO3
Molecular Weight (g/mol) 212.017
MDL Number MFCD22124929
InChI Key XQUUVPNLRZQWNF-UHFFFAOYSA-N
Synonym Potassium (Trifluoromethyl)trimethoxyborate
PubChem CID 23682739
IUPAC Name potassium;trimethoxy(trifluoromethyl)boranuide
SMILES [B-](C(F)(F)F)(OC)(OC)OC.[K+]

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 212.02
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Trimethoxy(trifluoromethyl)borate