missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Crystal, ACS, 98.5%, Spectrum™ Chemical

$250.09 - $13905.09

Chemical Identifiers

CAS 333-20-0
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612927
View Documents
Spectrum Chemical Mfg Cor
P142025GM
25 g Poly Bottle
Each for $250.09
Only null left
 
18612928
View Documents
Spectrum Chemical Mfg Cor
P1420125GM
125 g Poly Bottle
Each for $372.06
Only null left
 
18612929
View Documents
Spectrum Chemical Mfg Cor
P1420500GM
500 g Poly Bottle
Each for $875.26
Only null left
 
18612930
View Documents
Spectrum Chemical Mfg Cor
P142025KG
2.5 kg Poly Bottle
Each for $2,734.32
Only null left
 
18612931
View Documents
Spectrum Chemical Mfg Cor
P142012KG
12 kg Poly Pail
Each for $4,859.41
Only null left
 
18612932
View Documents
Spectrum Chemical Mfg Cor
P142050KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

333-20-0

Specifications

333-20-0
5.3 to 8.7
98.5%
Poly Bottle
1
25 g
ACS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.