missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Crystal, ACS, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P142012KG

 View more versions of this product

Catalog No. 18612931


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

333-20-0
5.3 to 8.7
98.5%
Poly Pail
1
12 kg
ACS

Chemical Identifiers

333-20-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.