missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, Crystal, ACS, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| pH | 5.3 to 8.7 |
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 50 kg |
| Percent Purity | 98.5% |
| Grade | ACS/Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Thiocyanate, Crystal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.