missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, 0.100 N (N/10), Ricca Chemical

Catalog No. p-3252209
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
658016 500 mL Natural Poly Bottle
658032 1 L Natural Poly Bottle
65801 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 658016 Supplier Ricca Chemical Company Supplier No. 658016
Only null left

Chemical Identifiers

CAS 333-20-0 , 7732-18-5
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Color Colorless
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Grade Laboratory
Packaging Natural Poly Bottle
pH Neutral
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.100N
Chemical Name or Material Potassium Thiocyanate