missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, 0.100 N (N/10), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 97.0, 0.97 |
| CAS Max % | 100.0, 1.01 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | Neutral |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.100N |
| Chemical Name or Material | Potassium Thiocyanate |