missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company 65801
Specifications
| Potassium Thiocyanate | |
| 333-20-0,7732-18-5 | |
| 100.0,1.01 | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| C(#N)[S-].[K+] | |
| 97.176 | |
| CHEBI:30951 | |
| Natural Poly Bottle | |
| 4 L | |
| 0.100N |
| Colorless | |
| 97.0,0.97 | |
| CKNS | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| potassium;thiocyanate | |
| 516872 | |
| Laboratory | |
| Neutral | |
| Liquid |
Chemical Identifiers
| 333-20-0 | |
| 97.176 | |
| potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
| CHEBI:30951 | |
| C(#N)[S-].[K+] |
| CKNS | |
| ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
| 516872 | |
| potassium;thiocyanate |