missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Potassium tert-butoxide solution

Numéro de catalogue. 328650500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
328650500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 328650500ML Fournisseur Sigma Aldrich Code fournisseur 328650500ML
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium tert-butoxide solution is a strong non nucleophilic base. It is capable of deprotonating carbon and other Bronsted acids. Reagent solutions in tert-BuOh may be prepared by reacting anhydrous alcohol with potassium in nitrogen atmosphere.Potassium tert-butoxide is used to prepare 3-potassiooxamethylpyridine catalyst. It is also used as a strong alkoxide base reagent.

Spécifications

CAS 865-47-4
Density 0.902 g/mL (at 25°C)
Molecular Formula C4H9KO
Linear Formula (CH3)3 COK
Quantity 500 mL
MDL Number MFCD00012162
Synonym Potassium tert-butylate; Potassium-2-methylpropan-2-olate solution
Molecular Weight (g/mol) 112.21
Biological Activity Respiratory system