missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tellurite, MP Biomedicals

Catalog No. 0210042780
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0210042780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0210042780 Supplier MP Biomedicals Inc Supplier No. 0210042780
Only null left

Chemical Identifiers

CAS 7790-58-1
Molecular Formula K2O3Te
Molecular Weight (g/mol) 253.794
InChI Key BFPJYWDBBLZXOM-UHFFFAOYSA-L
Synonym potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e
PubChem CID 65186
ChEBI CHEBI:75248
IUPAC Name dipotassium;tellurite
SMILES [O-][Te](=O)[O-].[K+].[K+]

Specifications

Quantity 100 g
Formula Weight 253.8
Chemical Name or Material Potassium Tellurite
Recommended Storage Room Temperature