missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyroantimonate TS, Ricca Chemical

Catalog No. 645832
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
645832 1 L Natural Poly Bottle
645816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 645832 Supplier Ricca Chemical Company Supplier No. 645832
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3 , 12208-13-8 , 1310-73-2
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color Colorless
CAS Min % 95.0, 1.56, 1.25, 0.1
CAS Max % 99.0, 1.63, 1.3, 0.11
Grade Laboratory
Packaging Natural Poly Bottle
pH >12
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Pyroantimonate, Test Solution