missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pivalate 98.0+%, TCI America™

Catalog No. P23545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23545G 5 g
P235425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23545G Supplier TCI America Supplier No. P23545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19455-23-3
Molecular Formula C5H9KO2
Molecular Weight (g/mol) 140.223
MDL Number MFCD00671345
InChI Key WFMNHCSATCWAAQ-UHFFFAOYSA-M
Synonym potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt
PubChem CID 23662159
IUPAC Name potassium;2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)[O-].[K+]

Specifications

Melting Point 300°C
Color White
Quantity 5 g
Formula Weight 140.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Pivalate