missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, for HPLC, Honeywell Fluka™

Catalog No. 6002771
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Catalog No. Quantity
6002771 50 g
6002770 250 g
6002769 1 kg
3 options

Catalog No. 6002771

Supplier: Honeywell Chemicals 6022150G

Only null left

for HPLC,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Quantity 50 g
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Formula Weight 136.09g/mol
Percent Purity ≥99.5%
Density 2.338g/mL at 25°C
Chemical Name or Material Potassium phosphate monobasic
RTECSNumber TC6615500