missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, 1.0M solution in water

Numéro de catalogue. AC397080010
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC397080010 1 L Glass bottle
AC397080025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC397080010 Fournisseur Thermo Scientific Chemicals Code fournisseur 397080010
Il en reste null

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Chemical Name or Material Potassium phosphate, dibasic
CAS 7758-11-4, 7732-18-5
Quantity 1 L
Molecular Formula HK2O4P
Linear Formula K2HPO4
MDL Number MFCD00011383
Merck Index 15, 7779
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.18
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18
Concentration or Composition (by Analyte or Components) 1.0M, exact strength on the certificate of analysis
Packaging Glass bottle
Appearance Clear colorless solution
Specific Gravity 1.13
Density 1.1300g/mL
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only