missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium phosphate, 0.2M buffer soln., pH 8.0

Catalog No. AAJ60851AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60851AP 500 mL
AAJ60851K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60851AP Supplier Thermo Scientific Chemicals Supplier No. J60851AP
Only null left

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material Potassium phosphate
Recommended Storage Ambient temperatures

RUO – Research Use Only