missing translation for 'onlineSavingsMsg'
Learn More

Potassium phenyltrifluoroborate, 98%

Catalog No. AAL1756803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1756803 1 g
AAL1756806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1756803 Supplier Thermo Scientific Chemicals Supplier No. L1756803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 153766-81-5 | C6H5BF3K | 184.01 g/mol

Aryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of Dicarbonyl­(2,4-pentanedionato)­rhodium(I)­, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aryltrifluoroborate salts are more nucleophilic than the corresponding arylboronic acids and undergo ligand-free Pd-catalyzed cross-coupling reactions with arenediazonium tetrafluoroborates to give good yields of biaryls. Suzuki Cross-Coupling are conducted using organotrifluoroborates as a potent boronic acid surrogates. In the presence of Dicarbonyl­(2,4-pentanedionato)­rhodium(I)­, 39295, K aryl- and alkenyltrifluoroborates add to aldehydes and enones to give secondary alcohols and saturated ketones, respectively.

Solubility
Soluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 153766-81-5
Molecular Formula C6H5BF3K
Molecular Weight (g/mol) 184.01
MDL Number MFCD01318172
InChI Key DVAFPKUGAUFBTJ-UHFFFAOYSA-N
Synonym potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide
PubChem CID 23675248
IUPAC Name potassium;trifluoro(phenyl)boranuide
SMILES [B-](C1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Melting Point 296°C to 301°C
Quantity 1 g
Beilstein 7782070
Solubility Information Soluble in water.
Formula Weight 184
Percent Purity 98%
Chemical Name or Material Potassium phenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only