missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate Solution, 1N (Certified), Fisher Chemical™

Catalog No. SP2824
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP2824 4 L Glass Bottle
SP2821 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP2824 Supplier SCP Science Supplier No. SP2824

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
Boiling Point 100°C
Melting Point 0°C
Concentration 0.995 to 1.005N
Density 1.0g/cm³
Grade Certified
Concentration or Composition (by Analyte or Components) 1N
Identification Pass Test
Packaging Glass Bottle
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Potassium Permanganate Solution, 1N
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1, : Oxidizer