accessibility menu, dialog, popup

UserName

CAS RN 7722-64-7

IUPAC Name: potassium manganesoylolate
Synonyms: potassium permanganate 高锰酸钾
Molecular Weight (g/mol): 158.03
Molecular Formula: KMnO4
InChi Key: VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES: [K+].[O-][Mn](=O)(=O)=O
Grade 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results