missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate Solution, 0.1N (N/10) (Certified), Fisher Chemical™

Catalog No. SP2881
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP2881 1 L Glass Bottle
SP2884 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP2881 Supplier SCP Science Supplier No. SP288 1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Titrated at 25°C against NIST™ sodium oxalate

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 7722-64-7 , 7732-18-5
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Purple
Boiling Point 100°C
Melting Point 0°C
Concentration 0.0995 to 0.1005N
Density 1.0g/cm³
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N (N/10)
Identification Pass Test
Packaging Glass Bottle
Traceability to NIST LOT NUMBER USED SODIUM OXALATE
Vapor Pressure 14mmHg
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Permanganate Solution, 0.1N (N/10)
Show More Show Less