missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oleate 98.0+%, TCI America™

Catalog No. O0056500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0056500G 500 g
O005625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0056500G Supplier TCI America Supplier No. O0056500G

Chemical Identifiers

CAS 143-18-0
Molecular Formula C18H33KO2
Molecular Weight (g/mol) 320.558
MDL Number MFCD00064243
InChI Key MLICVSDCCDDWMD-UHFFFAOYSA-M
Synonym acmc-209cpw, 9-octadecenoic acid 9z-, potassium salt 1:1, ksc175g6j
PubChem CID 44135690
IUPAC Name potassium;octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)[O-].[K+]

Specifications

Color White
Quantity 500 g
Formula Weight 320.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Oleate
EINECSNumber (2)-0611
RTECSNumber RK1150000
TSCA Yes