missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Crystal, Technical, Spectrum™ Chemical

$753.36 - $6,232.11

Chemical Identifiers

CAS 7758-09-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612809
View Documents
Spectrum Chemical Mfg Cor
P1354500GM
500 g Amber Glass Bottle N/A
Only null left
 
18612810
View Documents
Spectrum Chemical Mfg Cor
P135425KG
2.5 kg Poly Bottle N/A
Only null left
 
18612811
View Documents
Spectrum Chemical Mfg Cor
P135412KG
12 kg Poly Pail N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7758-09-0

Specifications

7758-09-0
500 g
Amber Glass Bottle
1
Technical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.