missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Crystal, Technical, Spectrum™ Chemical

Catalog No. 18612809
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612809 500 g Amber Glass Bottle
18612810 2.5 kg Poly Bottle
18612811 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612809 Supplier Spectrum Chemical Mfg Cor Supplier No. P1354500GM

Chemical Identifiers

CAS 7758-09-0

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.