missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Crystal, ACS, 96%, Spectrum™ Chemical

$467.81 - $31332.03

Chemical Identifiers

CAS 7758-09-0
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612804
View Documents
Spectrum Chemical Mfg Cor
P135325GM
25 g Poly Bottle
Each for $467.81
Only null left
 
18612805
View Documents
Spectrum Chemical Mfg Cor
P1353125GM
125 g Poly Bottle
Each for $671.94
Only null left
 
18612806
View Documents
Spectrum Chemical Mfg Cor
P1353500GM
500 g Poly Bottle
Each for $1,560.50
Only null left
 
18612807
View Documents
Spectrum Chemical Mfg Cor
P135325KG
2.5 kg Poly Bottle
Each for $4,960.03
Only null left
 
18612808
View Documents
Spectrum Chemical Mfg Cor
P135325KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7758-09-0

Specifications

7758-09-0
25 g
ACS
1
96%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.