missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrite, Crystal, ACS, 96%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P135325KG

 View more versions of this product

Catalog No. 18612807


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7758-09-0
2.5 kg
ACS
1
96%
Poly Bottle

Chemical Identifiers

7758-09-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.