missing translation for 'onlineSavingsMsg'
Learn More

Potassium methoxide, pure, 0.1 N solution in Toluene/Methanol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 240605000

 View more versions of this product

Catalog No. AC240605000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium methoxide
865-33-8,67-56-1,108-88-3
CH3OK
0.8500g/mL
Solubility in water: may decompose.
C[O-].[K+]
70.132
70.13
Glass bottle
500 mL
Liquid
Colorless
CH3KO
0.1N, exact strength on the certificate of analysis
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
BDAWXSQJJCIFIK-UHFFFAOYSA-N
potassium;methanolate
23664618
Pure
0.85
7°C

Chemical Identifiers

865-33-8
70.132
potassium methoxide, potassium methylate, potassium methanolate, methanol, potassium salt, unii-45myq0gwgb, 45myq0gwgb, potassiummethoxide, methanol, potassium salt 1:1, kome, meok
potassium;methanolate
CH3KO
BDAWXSQJJCIFIK-UHFFFAOYSA-N
23664618
C[O-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only