missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide USP/FCC Powder, Macron Fine Chemicals™

Catalog No. MK1115500
Change view
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly
POLYSTORMOR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK1115500 500g POLYSTORMOR
MK1115212 2.5kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK1115500 Supplier Avantor Supplier No. 111504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 500g
Formula Weight 166
Grade FCC/USP
Packaging POLYSTORMOR
Chemical Name or Material Potassium Iodide