Learn More
Potassium Iodide Solution (10% W/v), Fisher Chemical™
Supplier: Fisher Chemical SP2424
Specifications
| Potassium Iodide Solution | |
| 10% (w/v) | |
| 10% w/v | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 10% w/v | |
| Glass Bottle | |
| 4 L |
| Undesignated | |
| 7681-11-0 , 7732-18-5 | |
| IK | |
| 1.032g/cm³ | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 9 to 11% | |
| Pass Test | |
| 14mmHg | |
| Liquid |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0