missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015467
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015467 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015467 Supplier Merck Emd Millipore Supplier No. PHR13605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Iodide is well-known for being the first drug in the successful treatment of human sporotrichosis. Today it is widely used for the treatment of various kinds of skin inflammation disorders such as erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet′s syndrome. On account of its low cost, it continues to be used for cutaneous and lymphocutaneous diseases. It is also a part of the electrolyte of dye sensitized solar cells (DSSC).Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 7681-11-0
Vapor Pressure 1 mmHg (745°C)
Physical Form Neat
Linear Formula KI
MDL Number MFCD00011405
Melting Point 681°C
Synonym Potassium iodide
Type Certified Reference Material
Quantity 5 g
Formula Weight 166.00
Grade Certified Reference Material
Show More Show Less