missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodure de potassium (granulaire, à flux libre/USP/FCC), Fisher Chemical™
Spécifications
Spécifications
| Point d’ébullition | 1330 °C |
| Nom ou substance chimique | Potassium Iodide |
| Point de fusion | 680 °C |
| Couleur | Blanc |
| pH | 6 à 8 ans |
| Forme physique | Solide |
| Quantité | 500 g |
| Poids de formule | 166 |
| Pourcentage de pureté | 99,0 à 101,5% (USP) |
| Niveau | FCC/USP |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:1 Flammability:1 Instability:0 |