missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 1.0N, Fisher Chemical™

Catalog No. p-213916
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP8211 1 L Glass Bottle
SP8214 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP8211 Supplier Thermo Fisher Scientific Supplier No. SP8211
Only null left

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Undesignated
Concentration 0.995 to 1.005N
Density 1.032g/cm³
Concentration or Composition (by Analyte or Components) 1.0N
Packaging Glass Bottle
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Iodide, 1.0N