missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 0.33 N (0.33 M), Ricca Chemical

Supplier:  Ricca Chemical Company R63163004B

Quantity 4 L
Packaging Black Poly Bottle

 View more versions of this product

Catalog No. R63163004B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7681-11-0 , 144-55-8 , 7732-18-5
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide

Specifications

Potassium Iodide
7681-11-0 , 144-55-8 , 7732-18-5
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
Odorless
0.330 Normal, Stabilized
4 L
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
[K+].[I-]
166.003
CHEBI:8346
Black Poly Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15°C - 30°C (59°F - 86°F)

SDS
Documents

Documents

Promotions

Promotions