missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, 0.33 N (0.33 M), Ricca Chemical
Supplier: Ricca Chemical Company R63163001B
| Quantity | 1 L |
|---|---|
| Packaging | Amber Poly Bottle |
Chemical Identifiers
| 7681-11-0 , 144-55-8 , 7732-18-5 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Specifications
| Potassium Iodide | |
| 7681-11-0 , 144-55-8 , 7732-18-5 | |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| Odorless |
| 0.330 Normal, Stabilized | |
| 1 L | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| Amber Poly Bottle |
Safety and Handling
Recommended Storage : 15°C - 30°C (59°F - 86°F)