missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 0.100 Molar (M/10), 0.100 Normal (N/10), Stabilized, Ricca Chemical

Catalog No. 01213124
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01213124 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01213124 Supplier Ricca Chemical Company Supplier No. R63160001C
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7681-11-0 , 144-55-8 , 7681-11-0 , 7732-18-5
Molecular Weight (g/mol) 166 g/mol

Specifications

Boiling Point 100°C
Melting Point 0.0°C
Color Colorless
CAS Min % 98.35%, 1.64%
CAS Max % 98.35%, 1.64%
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Packaging 1 L Amber Glass Bottle
Appearance Colorless Liquid
Chemical Name or Material Potassium Iodide
Show More Show Less
ShelfLife 12 Months